Product Details

SNP ID
rs9487650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111492249 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAATTACATATCCAGAATTGATA[C/T]TGGAGCATTTCCAATGGATTATATT
Phenotype
MIM: 602776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REV3L PubMed Links

Gene Details

Gene
REV3L
Gene Name
REV3 like, DNA directed polymerase zeta catalytic subunit
There are no transcripts associated with this gene.

Gene
TRAF3IP2-AS1
Gene Name
TRAF3IP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details