Product Details

SNP ID
rs9979060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42900965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTGAATCCAGTACCCAGGCCT[A/G]ATTTTGCAACAGTCAGTAATAAATT
Phenotype
MIM: 602184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFV3 PubMed Links

Gene Details

Gene
NDUFV3
Gene Name
NADH:ubiquinone oxidoreductase subunit V3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001503.1 Intron NP_001001503.1
NM_021075.3 Intron NP_066553.3
XM_011529586.2 Intron XP_011527888.1
XM_017028359.1 Intron XP_016883848.1

View Full Product Details