Product Details
- SNP ID
-
rs4951301
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:204080085 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTGTGGCATAGTAGAGGAAAAGA[A/G]GGAAGAAGTGGGGACAGTATCTCCA
- Phenotype
-
MIM: 604748
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SOX13
PubMed Links
Gene Details
- Gene
- SOX13
- Gene Name
- SRY-box 13
View Full Product Details