Product Details

SNP ID
rs9973589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101253306 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGTGCTCGTCATGCTGCTCCAGCA[A/G]CCCGACCTGCTGCCTAGCGCGGCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017546.4 424 Silent Mutation CAA,CAG Q114Q NP_060016.3

View Full Product Details