Product Details

SNP ID
rs6121044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35778822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACAACTTTGCTAGATCTTGGG[A/C]TAGGTCCTACCTGGACCTTCCTCTC
Phenotype
MIM: 610335
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PHF20 PubMed Links

Gene Details

Gene
PHF20
Gene Name
PHD finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016436.4 Intron NP_057520.2
XM_005260421.2 Intron XP_005260478.1
XM_011528843.1 Intron XP_011527145.1
XM_011528844.1 Intron XP_011527146.1
XM_017027864.1 Intron XP_016883353.1
XM_017027865.1 Intron XP_016883354.1
XM_017027866.1 Intron XP_016883355.1
XM_017027867.1 Intron XP_016883356.1
XM_017027868.1 Intron XP_016883357.1
XM_017027869.1 Intron XP_016883358.1
XM_017027870.1 Intron XP_016883359.1
XM_017027871.1 Intron XP_016883360.1

View Full Product Details