Product Details
- SNP ID
-
hCV30092851
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:114235094 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGGCCCGCGCCGCCATGAACCTA[G/T]AGCGGCTGCGGAAGCGCGTCCGGCA
- Phenotype
-
MIM: 603461
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDC16
PubMed Links
Gene Details
- Gene
- CDC16
- Gene Name
- cell division cycle 16
There are no transcripts associated with this gene.
- Gene
- MIR548AR
- Gene Name
- microRNA 548ar
There are no transcripts associated with this gene.
View Full Product Details