Product Details
- SNP ID
-
rs10447099
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:62417350 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTTTTTTTTTTTTTTTTTTTGG[G/T]ATTTTGAAGGCTGACAGTCCATAGC
- Phenotype
-
MIM: 610889
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IPO11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76466551] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IPO11
- Gene Name
- importin 11
View Full Product Details