Product Details

SNP ID
rs6679068
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:46860554 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCAACAAGGTCAGCAATGCTAGA[C/T]ATTGGTTCATTGAAAAGATGAATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4Z2P PubMed Links

Gene Details

Gene
CYP4Z2P
Gene Name
cytochrome P450 family 4 subfamily Z member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details