Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000873.3 | 2359 | Intron | NP_000864.2 | ||
NM_001099786.1 | 2359 | Intron | NP_001093256.1 | ||
NM_001099787.1 | 2359 | Intron | NP_001093257.1 | ||
NM_001099788.1 | 2359 | Intron | NP_001093258.1 | ||
NM_001099789.1 | 2359 | Intron | NP_001093259.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164257.1 | 2359 | UTR 3 | NP_001157729.1 | ||
NM_001191029.1 | 2359 | UTR 3 | NP_001177958.1 | ||
NM_001191030.1 | 2359 | UTR 3 | NP_001177959.1 | ||
NM_001191031.1 | 2359 | UTR 3 | NP_001177960.1 | ||
XM_005257805.4 | 2359 | Intron | XP_005257862.1 | ||
XM_011525468.2 | 2359 | Intron | XP_011523770.1 | ||
XM_011525469.1 | 2359 | UTR 3 | XP_011523771.1 | ||
XM_017025315.1 | 2359 | UTR 3 | XP_016880804.1 |