Product Details

SNP ID
rs7799136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138591889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTAGAGGCATACGACAACGATTG[C/T]GGTTTCAGTTCCAGACCATTGCAAT
Phenotype
MIM: 611700 MIM: 603406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SVOPL PubMed Links

Gene Details

Gene
SVOPL
Gene Name
SVOP like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139456.1 3891 Intron NP_001132928.1
NM_174959.3 3891 Intron NP_777619.1
XM_005250143.3 3891 UTR 3 XP_005250200.1
XM_011515797.2 3891 UTR 3 XP_011514099.1
XM_017011746.1 3891 UTR 3 XP_016867235.1
XM_017011747.1 3891 UTR 3 XP_016867236.1
XM_017011748.1 3891 UTR 3 XP_016867237.1
Gene
TRIM24
Gene Name
tripartite motif containing 24
There are no transcripts associated with this gene.

View Full Product Details