Product Details

SNP ID
rs10516674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82437609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAGTAAGAGCTACCTCTTCTTG[C/T]AAAGGACCTATTTAAGCAGGAATTT
Phenotype
MIM: 607137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENOPH1 PubMed Links

Gene Details

Gene
ENOPH1
Gene Name
enolase-phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292017.1 Intron NP_001278946.1
NM_021204.4 Intron NP_067027.1
Gene
HNRNPDL
Gene Name
heterogeneous nuclear ribonucleoprotein D like
There are no transcripts associated with this gene.

View Full Product Details