Product Details

SNP ID
rs9855205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45894206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGAGATTTATTTCAGTCAGTTC[C/T]GAGGCCCTGTGCATTAGGGAACTAG
Phenotype
MIM: 604738 MIM: 606568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR9 PubMed Links

Gene Details

Gene
CCR9
Gene Name
C-C motif chemokine receptor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256369.1 770 Intron NP_001243298.1
NM_006641.3 770 Intron NP_006632.2
NM_031200.2 770 Intron NP_112477.1
XM_011533312.2 770 UTR 5 XP_011531614.1
Gene
LZTFL1
Gene Name
leucine zipper transcription factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276378.1 770 Intron NP_001263307.1
NM_001276379.1 770 Intron NP_001263308.1
NM_020347.3 770 Intron NP_065080.1
XM_006713207.2 770 Intron XP_006713270.2
XM_011533838.2 770 Intron XP_011532140.1
XM_017006644.1 770 Intron XP_016862133.1
XM_017006645.1 770 Intron XP_016862134.1

View Full Product Details