Product Details

SNP ID
rs10068033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:156852012 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCCTACTCCTTTCCACACATA[C/T]ACACACAATCTTTTACCTTTTAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R2P3 PubMed Links

Gene Details

Gene
PPP1R2P3
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details