Product Details

SNP ID
rs9464014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54019844 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAATACATTGTTCTAGATGTAT[A/G]AAGTTGAGGTTTATGGACTATTCAA
Phenotype
MIM: 614106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLIP PubMed Links

Gene Details

Gene
MLIP
Gene Name
muscular LMNA-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281746.1 Intron NP_001268675.1
NM_001281747.1 Intron NP_001268676.1
NM_138569.2 Intron NP_612636.2
XM_005249476.4 Intron XP_005249533.1
XM_005249478.4 Intron XP_005249535.1
XM_005249480.4 Intron XP_005249537.1
XM_006715245.3 Intron XP_006715308.2
XM_017011465.1 Intron XP_016866954.1
XM_017011466.1 Intron XP_016866955.1
XM_017011467.1 Intron XP_016866956.1
XM_017011468.1 Intron XP_016866957.1
XM_017011469.1 Intron XP_016866958.1
XM_017011470.1 Intron XP_016866959.1

View Full Product Details