Product Details

SNP ID
rs6067471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50510745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAGCGACGCACTGCGTCCCCCC[A/C]CCCTTTGTCCCCGGGGCGGGCGTGT
Phenotype
MIM: 176885
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTPN1 PubMed Links

Gene Details

Gene
PTPN1
Gene Name
protein tyrosine phosphatase, non-receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278618.1 Intron NP_001265547.1
NM_002827.3 Intron NP_002818.1

View Full Product Details