Product Details

SNP ID
rs4968005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23467214 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGCACAGAGTGGATGAATGGAAC[A/G]AACGATACATGAAAAAAACACTTTG
Phenotype
MIM: 606005
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGA2 PubMed Links

Gene Details

Gene
GGA2
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015044.4 2304 UTR 3 NP_055859.1
XM_017023074.1 2304 UTR 3 XP_016878563.1
XM_017023075.1 2304 UTR 3 XP_016878564.1

View Full Product Details