Product Details

SNP ID
rs7524203
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:212838470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCTGGAATTCTCATATGCTGCTA[C/G]TGAGAATGTAAAATGGAACAATGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPATA45 PubMed Links

Gene Details

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 Intron NP_001019772.1

View Full Product Details