Product Details
- SNP ID
-
rs9894033
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41459566 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATTCTCAATTACAGTAATTTGG[C/T]CTAGGTTTCTGGCATAATTATCTCC
- Phenotype
-
MIM: 602760
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT32
PubMed Links
Gene Details
- Gene
- KRT32
- Gene Name
- keratin 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002278.3 |
1952 |
UTR 3 |
|
|
NP_002269.3 |
View Full Product Details