Product Details
- SNP ID
-
rs6068888
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:54478356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGAATGTTAGCGGTATTAATCTA[A/G]GCTACATATAGTCTAGAACAGCATG
- Phenotype
-
MIM: 608334
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DOK5
PubMed Links
Gene Details
- Gene
- DOK5
- Gene Name
- docking protein 5
View Full Product Details