Product Details
- SNP ID
-
rs6114205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23629316 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACGCTGGCCGCTGGCCTCAGAGGT[A/G]TGTTTGGTAATGATGCCTCCATGCA
- Phenotype
-
MIM: 604312
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST3
PubMed Links
Gene Details
- Gene
- CST3
- Gene Name
- cystatin C
View Full Product Details