Product Details

SNP ID
rs9965456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:14076991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCAATTGGTGAACTCTTACACA[C/T]TATCGAAGGCATGAAGGTGTTGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF519 PubMed Links

Gene Details

Gene
ZNF519
Gene Name
zinc finger protein 519
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145287.3 10713 Intron NP_660330.2
XM_011525605.2 10713 Intron XP_011523907.1
XM_017025562.1 10713 UTR 3 XP_016881051.1
XM_017025563.1 10713 UTR 3 XP_016881052.1
XM_017025564.1 10713 UTR 3 XP_016881053.1
XM_017025565.1 10713 UTR 3 XP_016881054.1
XM_017025566.1 10713 Intron XP_016881055.1

View Full Product Details