Product Details
- SNP ID
-
rs10135990
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:95701719 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGCCAGTTGAGTGAGAAGAGGGA[A/G]AAATTTCTCGAACCCATCTCCCTTT
- Phenotype
-
MIM: 186960
MIM: 603769
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TCL1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113631765] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCL1A
- Gene Name
- T-cell leukemia/lymphoma 1A
- Gene
- TCL1B
- Gene Name
- T-cell leukemia/lymphoma 1B
There are no transcripts associated with this gene.
View Full Product Details