Product Details

SNP ID
rs8100597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52936940 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCTGACCGCCGGTCCTCTGGGC[C/T]CACTTTTCTCCCTCTGTACTTTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF321P PubMed Links

Gene Details

Gene
ZNF321P
Gene Name
zinc finger protein 321, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF816-ZNF321P
Gene Name
ZNF816-ZNF321P readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202473.1 Intron NP_001189402.1
Gene
ZNF888
Gene Name
zinc finger protein 888
There are no transcripts associated with this gene.

View Full Product Details