Product Details
- SNP ID
-
rs6115976
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:435556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACAGCCAGATGAAGATACACACA[A/G]GGTGAGGTCTGGACGGGTCCTGGGC
- Phenotype
-
MIM: 610924
MIM: 611663
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RBCK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs375424312] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RBCK1
- Gene Name
- RANBP2-type and C3HC4-type zinc finger containing 1
There are no transcripts associated with this gene.
- Gene
- TBC1D20
- Gene Name
- TBC1 domain family member 20
View Full Product Details