Product Details

SNP ID
rs10123412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93589198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCTGGGAAGGAACCAAGTGCTC[A/G]CTGAAAGTTTGGAGCAGGAAGCTTT
Phenotype
MIM: 604351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR548AU PubMed Links

Gene Details

Gene
MIR548AU
Gene Name
microRNA 548au
There are no transcripts associated with this gene.

Gene
PHF2
Gene Name
PHD finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005392.3 Intron NP_005383.3
XM_005252051.2 Intron XP_005252108.1
XM_006717143.2 Intron XP_006717206.1

View Full Product Details