Product Details

SNP ID
rs6057766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33220443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTGGATTGGAGTTAAAATGAAT[G/T]TGTTAATTTAAAGAGAGTTGATATT
Phenotype
MIM: 607627
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BPIFA3 PubMed Links

Gene Details

Gene
BPIFA3
Gene Name
BPI fold containing family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042439.2 Intron NP_001035904.1
NM_178466.4 Intron NP_848561.2
Gene
BPIFA4P
Gene Name
BPI fold containing family A member 4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details