Product Details

SNP ID
rs10454626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51357017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATGGCCTACCACCTGCTTTTGT[A/G]AAGTTTAATTAAAACATAGCCACTC
Phenotype
MIM: 604331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INTS6 PubMed Links

Gene Details

Gene
INTS6
Gene Name
integrator complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039937.1 Intron NP_001035026.1
NM_001039938.1 Intron NP_001035027.1
NM_001306091.1 Intron NP_001293020.1
NM_012141.2 Intron NP_036273.1
XM_011535040.2 Intron XP_011533342.1
XM_011535041.1 Intron XP_011533343.1
XM_017020540.1 Intron XP_016876029.1
Gene
MIR5693
Gene Name
microRNA 5693
There are no transcripts associated with this gene.

Gene
SERPINE3
Gene Name
serpin family E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101320.1 Intron NP_001094790.1
XM_017020707.1 Intron XP_016876196.1

View Full Product Details