Product Details
- SNP ID
-
rs9822116
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179356827 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGGACCCAAACTGTAACTATAG[G/T]AGGTCCATCACCCAATGTATGCAGC
- Phenotype
-
MIM: 608506
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MFN1
PubMed Links
Gene Details
- Gene
- MFN1
- Gene Name
- mitofusin 1
View Full Product Details