Product Details

SNP ID
rs9941771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:24953889 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTTCTTTCCGTTGACGTCATCG[C/T]GGCCATCACCACCATCACCCTCTGC
Phenotype
MIM: 615884 MIM: 603253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APMAP PubMed Links

Gene Details

Gene
APMAP
Gene Name
adipocyte plasma membrane associated protein
There are no transcripts associated with this gene.

Gene
CST7
Gene Name
cystatin F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003650.3 Intron NP_003641.3

View Full Product Details