Product Details
- SNP ID
-
rs9324068
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103935613 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTAGACACTGGGAAGTTTAACTT[C/T]GAGAATGATGAGATCCCTTAACTAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RD3L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75340877] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RD3L
- Gene Name
- retinal degeneration 3-like
There are no transcripts associated with this gene.
- Gene
- TDRD9
- Gene Name
- tudor domain containing 9
View Full Product Details