Product Details

SNP ID
rs8104891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53872868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATGGCTGAGCTACTGAGTCCCT[G/T]GGGGAGGGAGGGAAAGAGGGAAGCA
Phenotype
MIM: 609959 MIM: 176980
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372457 PubMed Links

Gene Details

Gene
LOC105372457
Gene Name
uncharacterized LOC105372457
There are no transcripts associated with this gene.

Gene
MYADM
Gene Name
myeloid associated differentiation marker
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001020818.2 Intron NP_001018654.1
NM_001020819.2 Intron NP_001018655.1
NM_001020820.2 Intron NP_001018656.1
NM_001020821.2 Intron NP_001018657.1
NM_001290188.1 Intron NP_001277117.1
NM_001290189.1 Intron NP_001277118.1
NM_001290190.1 Intron NP_001277119.1
NM_001290191.1 Intron NP_001277120.1
NM_001290192.1 Intron NP_001277121.1
NM_001290193.1 Intron NP_001277122.1
NM_001290194.1 Intron NP_001277123.1
NM_138373.4 Intron NP_612382.1
Gene
PRKCG
Gene Name
protein kinase C gamma
There are no transcripts associated with this gene.

View Full Product Details