Product Details
- SNP ID
-
rs9458302
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:157717630 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCCTCTGCAGCTCCTAACACCAA[C/G]GTCTTACGCACAGGAAGTGCTCCAT
- Phenotype
-
MIM: 605952
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SNX9
PubMed Links
Gene Details
- Gene
- SNX9
- Gene Name
- sorting nexin 9
View Full Product Details