Product Details

SNP ID
rs9943752
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:24997179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTATAATCAGGATGAGCTTTAG[A/T]GTGTTGTTTTTTGTAGAGCCGGGCG
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
C12orf77 PubMed Links

Gene Details

Gene
C12orf77
Gene Name
chromosome 12 open reading frame 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101339.2 Intron NP_001094809.2
XM_017018966.1 Intron XP_016874455.1
Gene
LOC645177
Gene Name
uncharacterized LOC645177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321724.1 Intron NP_001308653.1

View Full Product Details