Product Details
- SNP ID
-
rs9994632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:74366948 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAGTTCTGTATTATATCTCTAAA[C/T]TCTATTCAGTTATGCAGAGCCCATA
- Phenotype
-
MIM: 602061
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EREG
PubMed Links
Gene Details
- Gene
- EREG
- Gene Name
- epiregulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001432.2 |
|
Intron |
|
|
NP_001423.1 |
View Full Product Details