Product Details
- SNP ID
-
rs6695152
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
43
- Location
-
Chr.1:172538572 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTCTGCTTCTGTCTCTAAACTTT[A/C]AAGGTACGGAATCTATTTCTTTTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SUCO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78223831] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUCO
- Gene Name
- SUN domain containing ossification factor
View Full Product Details