Product Details

SNP ID
rs10246237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103101457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAAAATCAGTGAAAGTTTTCA[A/G]TGTTTAGTCTCCTAATATATAATGC
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
There are no transcripts associated with this gene.

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 3482 UTR 3 NP_001116310.1
NM_198990.4 3482 UTR 3 NP_945341.3
XM_005250214.4 3482 UTR 3 XP_005250271.2
XM_005250215.2 3482 UTR 3 XP_005250272.1
XM_005250218.2 3482 UTR 3 XP_005250275.1
XM_011515933.2 3482 UTR 3 XP_011514235.1
XM_011515934.2 3482 UTR 3 XP_011514236.1
XM_017011854.1 3482 UTR 3 XP_016867343.1
XM_017011855.1 3482 UTR 3 XP_016867344.1
XM_017011856.1 3482 Intron XP_016867345.1
XM_017011857.1 3482 Intron XP_016867346.1

View Full Product Details