Product Details

SNP ID
hCV30283790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37959655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCCCTCAGGGAGCTTGTGAT[C/T]TAATAGGAGAACTAGGACATGAACA
Phenotype
MIM: 604414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf23 PubMed Links

Gene Details

Gene
C22orf23
Gene Name
chromosome 22 open reading frame 23
There are no transcripts associated with this gene.

Gene
MIR6820
Gene Name
microRNA 6820
There are no transcripts associated with this gene.

Gene
POLR2F
Gene Name
RNA polymerase II subunit F
There are no transcripts associated with this gene.

View Full Product Details