Product Details

SNP ID
rs7549959
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:15241304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATCTCCCAAACATAACGTCAAGC[A/G]AAAGAAACCAGACAGAAGATGGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FHAD1 PubMed Links

Gene Details

Gene
FHAD1
Gene Name
forkhead associated phosphopeptide binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052929.1 Intron NP_443161.1
XM_011540575.2 Intron XP_011538877.1
XM_011540576.2 Intron XP_011538878.1
XM_011540577.2 Intron XP_011538879.1
XM_011540580.2 Intron XP_011538882.1
XM_011540581.2 Intron XP_011538883.1
XM_011540582.2 Intron XP_011538884.1
XM_011540584.2 Intron XP_011538886.1
XM_011540588.2 Intron XP_011538890.1
XM_011540589.2 Intron XP_011538891.1
XM_011540590.2 Intron XP_011538892.1
XM_011540592.2 Intron XP_011538894.1
XM_011540593.2 Intron XP_011538895.1
XM_011540595.2 Intron XP_011538897.1
XM_011540596.2 Intron XP_011538898.1
XM_011540597.2 Intron XP_011538899.1
XM_011540598.2 Intron XP_011538900.1
XM_017000198.1 Intron XP_016855687.1
XM_017000199.1 Intron XP_016855688.1
XM_017000200.1 Intron XP_016855689.1
XM_017000201.1 Intron XP_016855690.1
XM_017000202.1 Intron XP_016855691.1
XM_017000203.1 Intron XP_016855692.1
XM_017000204.1 Intron XP_016855693.1
XM_017000205.1 Intron XP_016855694.1
XM_017000206.1 Intron XP_016855695.1
XM_017000207.1 Intron XP_016855696.1
XM_017000208.1 Intron XP_016855697.1
XM_017000209.1 Intron XP_016855698.1
XM_017000210.1 Intron XP_016855699.1
XM_017000211.1 Intron XP_016855700.1
XM_017000212.1 Intron XP_016855701.1
XM_017000213.1 Intron XP_016855702.1
XM_017000214.1 Intron XP_016855703.1

View Full Product Details