Product Details

SNP ID
rs6609422
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46513355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCAATAACATATGAAATGATAAA[C/T]ACTTTCAATAGTAATCAGGGAAGTG
Phenotype
MIM: 300573
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 Intron NP_001034980.1
NM_001146291.1 Intron NP_001139763.1
NM_001190417.1 Intron NP_001177346.1
XM_011543941.2 Intron XP_011542243.1
XM_011543943.2 Intron XP_011542245.1
XM_011543944.2 Intron XP_011542246.1
XM_011543945.2 Intron XP_011542247.1
XM_017029728.1 Intron XP_016885217.1
XM_017029729.1 Intron XP_016885218.1
XM_017029730.1 Intron XP_016885219.1
XM_017029731.1 Intron XP_016885220.1
XM_017029732.1 Intron XP_016885221.1
XM_017029733.1 Intron XP_016885222.1

View Full Product Details