Product Details

SNP ID
rs10508861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:37775141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGTATCAGAGATAAGTCTGCAC[A/G]TTACTAATAATTACAGTTATACAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF248 PubMed Links

Gene Details

Gene
ZNF248
Gene Name
zinc finger protein 248
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267597.1 2288 Intron NP_001254526.1
NM_001267605.1 2288 Intron NP_001254534.1
NM_001267606.1 2288 Intron NP_001254535.1
NM_001267607.1 2288 Intron NP_001254536.1
NM_021045.2 2288 Intron NP_066383.1
XM_006717485.3 2288 Intron XP_006717548.1
XM_011519597.2 2288 Intron XP_011517899.1
XM_011519598.2 2288 Intron XP_011517900.1
XM_011519599.1 2288 Intron XP_011517901.1
XM_011519600.2 2288 Intron XP_011517902.1
XM_017016444.1 2288 Intron XP_016871933.1
XM_017016445.1 2288 Intron XP_016871934.1
XM_017016446.1 2288 Intron XP_016871935.1
XM_017016447.1 2288 Intron XP_016871936.1
XM_017016448.1 2288 Intron XP_016871937.1
XM_017016449.1 2288 UTR 3 XP_016871938.1
XM_017016450.1 2288 Intron XP_016871939.1
XM_017016451.1 2288 Intron XP_016871940.1

View Full Product Details