Product Details

SNP ID
rs121434240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1650898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGGCCTGTGCACCTCGTGGTA[G/C]AACTCTTTGGGGTTCGCCAGCTGTA
Phenotype
MIM: 607300 MIM: 607848 MIM: 607873
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
PRPF8 PubMed Links

Gene Details

Gene
PRPF8
Gene Name
pre-mRNA processing factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006445.3 7026 Silent Mutation TTC,TTG F2304L NP_006436.3
Gene
RILP
Gene Name
Rab interacting lysosomal protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031430.2 7026 Intron NP_113618.2
XM_005256811.1 7026 Intron XP_005256868.1
XM_017025195.1 7026 Intron XP_016880684.1
Gene
SCARF1
Gene Name
scavenger receptor class F member 1
There are no transcripts associated with this gene.

View Full Product Details