Product Details

SNP ID
rs9946054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2662792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTATAAGGTCATTTTTGTCTCT[G/T]TTAAACAAATAGAATCTTCCCAGTC
Phenotype
MIM: 614982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CBX3P2 PubMed Links

Gene Details

Gene
CBX3P2
Gene Name
chromobox 3 pseudogene 2
There are no transcripts associated with this gene.

Gene
SMCHD1
Gene Name
structural maintenance of chromosomes flexible hinge domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015295.2 Intron NP_056110.2
XM_011525642.1 Intron XP_011523944.1
XM_011525643.2 Intron XP_011523945.1
XM_011525644.2 Intron XP_011523946.1
XM_017025684.1 Intron XP_016881173.1

View Full Product Details