Product Details

SNP ID
rs6128751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:60049949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACCCTCTTGCTCCTCTCTCCC[A/C]TAGGGTCCTTGTCTCTCCTAGTTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C20orf197 PubMed Links

Gene Details

Gene
C20orf197
Gene Name
chromosome 20 open reading frame 197
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302813.1 Intron NP_001289742.1
NM_001302815.1 Intron NP_001289744.1
NM_173644.2 Intron NP_775915.1

View Full Product Details