Product Details

SNP ID
rs7705070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123527182 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTCATTTAGAAATAGTGATTTA[C/T]GGGAGCCTCATTTTTCAATGAATGC
Phenotype
MIM: 604253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSNK1G3 PubMed Links

Gene Details

Gene
CSNK1G3
Gene Name
casein kinase 1 gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031812.3 Intron NP_001026982.1
NM_001044723.2 Intron NP_001038188.1
NM_001270572.1 Intron NP_001257501.1
NM_001270573.1 Intron NP_001257502.1
NM_001270574.1 Intron NP_001257503.1
NM_004384.4 Intron NP_004375.2
XM_005271891.1 Intron XP_005271948.1
XM_005271892.2 Intron XP_005271949.1
XM_005271894.2 Intron XP_005271951.1
XM_005271895.2 Intron XP_005271952.1
XM_005271896.1 Intron XP_005271953.1
XM_017009057.1 Intron XP_016864546.1
XM_017009058.1 Intron XP_016864547.1
XM_017009059.1 Intron XP_016864548.1
XM_017009060.1 Intron XP_016864549.1
XM_017009061.1 Intron XP_016864550.1
XM_017009062.1 Intron XP_016864551.1
XM_017009063.1 Intron XP_016864552.1
XM_017009064.1 Intron XP_016864553.1
XM_017009065.1 Intron XP_016864554.1
XM_017009066.1 Intron XP_016864555.1
XM_017009067.1 Intron XP_016864556.1
XM_017009068.1 Intron XP_016864557.1
XM_017009069.1 Intron XP_016864558.1
XM_017009070.1 Intron XP_016864559.1

View Full Product Details