Product Details

SNP ID
rs6020280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37698249 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTGAAACCTCCTAAGTCTGCTA[C/T]TGGAGAGCACTTTGATCTCGCCAGC
Phenotype
MIM: 611537
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNBL1 PubMed Links

Gene Details

Gene
CTNNBL1
Gene Name
catenin beta like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281495.1 Intron NP_001268424.1
NM_030877.4 Intron NP_110517.2
XM_011528917.2 Intron XP_011527219.1

View Full Product Details