Product Details
- SNP ID
-
rs6071631
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:38970814 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACTATTCTTTGTCTCCCAACTGT[A/G]CTTACAAGATAACATAAATGAAACA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DHX35
PubMed Links
Gene Details
- Gene
- DHX35
- Gene Name
- DEAH-box helicase 35
View Full Product Details