Product Details

SNP ID
rs9372737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:123222304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGGTTCTAGAGCGGTTCCCCTCA[C/G]AGTGTAATCTATATACCACTAGTTT
Phenotype
MIM: 603283
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRDN PubMed Links

Gene Details

Gene
TRDN
Gene Name
triadin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251987.1 Intron NP_001238916.1
NM_001256020.1 Intron NP_001242949.1
NM_001256021.1 Intron NP_001242950.1
NM_001256022.1 Intron NP_001242951.1
NM_006073.3 Intron NP_006064.2

View Full Product Details