Product Details

SNP ID
rs7042382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19110469 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAAATTTTGACATTGGATTTC[G/T]TTTTGTTTTGTTTGTTTGTTTGTTT
Phenotype
MIM: 613433 MIM: 103195
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HAUS6 PubMed Links

Gene Details

Gene
HAUS6
Gene Name
HAUS augmin like complex subunit 6
There are no transcripts associated with this gene.

Gene
PLIN2
Gene Name
perilipin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122.3 Intron NP_001113.2
XM_017014259.1 Intron XP_016869748.1

View Full Product Details