Product Details
- SNP ID
-
rs9899063
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:31104389 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCAAGGGACAGGAGGTATCGGAA[A/G]GCTCTAAAGGAAGTTTAAGGAGGAG
- Phenotype
-
MIM: 613113
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4733
PubMed Links
Gene Details
- Gene
- MIR4733
- Gene Name
- microRNA 4733
There are no transcripts associated with this gene.
- Gene
- NF1
- Gene Name
- neurofibromin 1
View Full Product Details