Product Details

SNP ID
rs10506826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80424984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAGTTCTCCAAATGTGTCTGA[A/G]CTAAATGTTTTAATGTGTCACTTTT
Phenotype
MIM: 603317
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPRQ PubMed Links

Gene Details

Gene
PTPRQ
Gene Name
protein tyrosine phosphatase, receptor type Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145026.1 Intron NP_001138498.1
XM_017019273.1 Intron XP_016874762.1
XM_017019274.1 Intron XP_016874763.1
XM_017019275.1 Intron XP_016874764.1

View Full Product Details